Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitromethane-d{3}, 99%(Isotopic), Thermo Scientific Chemicals

Catalog No. AA4233806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4233806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4233806 Supplier Thermo Scientific Chemicals Supplier No. 04233806
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13031-32-8 | CH3NO2 | 64.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13031-32-8
Molecular Formula CH3NO2
Molecular Weight (g/mol) 64.06
MDL Number MFCD00044214
InChI Key LYGJENNIWJXYER-FIBGUPNXSA-N
Synonym nitromethane-d3, nitro 2h3 methane, nitro 2 h? methane, nitromethane-d3, 99 atom % d, methane-d3, nitro, cd3no2, trideuteronitromethane, methane-d3-, nitro, nitromethane-d3, ≥99 atom % d, contains 1 % v/v tms
PubChem CID 123293
IUPAC Name trideuterio(nitro)methane
SMILES [2H]C([2H])([2H])[N+]([O-])=O

Specifications

Physical Form Liquid
Quantity 5 g
Color Colorless
Density 1.183 g/mL
Refractive Index 1.3795
Odor Characteristic
Boiling Point 100°C
UN Number UN1261
Formula Weight 64.06
Percent Purity 99%
Chemical Name or Material Nitromethane-d3, Isotopic
Show More Show Less
Hazard Category H226-H302+H332-H351-H361
Hazard Statement GHS H Statement
H226-H302
Flammable liquid and vapor.
Harmful if swallowed.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P308+P313-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: NITROMETHANE
EINECSNumber 235-892-2
TSCA Yes
Recommended Storage Ambient

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.