Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitromethane-d3, for NMR, 99.5 atom % D

Catalog No. AC464230050
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464230050 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC464230050 Supplier Thermo Scientific Chemicals Supplier No. 464230050
Add to Cart

CAS: 13031-32-8 | CH3NO2 | 64.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13031-32-8
Molecular Formula CH3NO2
Molecular Weight (g/mol) 64.06
MDL Number MFCD00044214
InChI Key LYGJENNIWJXYER-FIBGUPNXSA-N
IUPAC Name nitro(²H₃)methane
SMILES [2H]C([2H])([2H])[N+]([O-])=O

Specifications

Density 1.1800 g/mL
Specific Gravity 1.18
Flash Point 35°C
Refractive Index 1.3795
Quantity 5 mL
Boiling Point 100.0°C to 102.0°C
Linear Formula CD3NO2
Formula Weight 64.06
Grade NMR
Chemical Name or Material Nitromethane-d3
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Flammable liquid and vapor.

Harmful if swallowed.

Harmful if inhaled.

Suspected of causing cancer.

Suspected of damaging fertility or the unborn child.

Precautionary Statement GHS P Statement Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.

No smoking.

IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Wash face, hands and any exposed skin thoroughly after handling.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

IF ON SKIN (or hair): Take off immediately all contaminated clothing.

Rinse skin with water or shower Wear protective gloves/protective clothing/eye protection/face protection.

EINECSNumber 235-892-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.