Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitromethane-d3, for NMR, 99 atom % D

Catalog No. AC166370250
Change view
Click to view available options
Quantity:
5 mL
10 mL
25 mL
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC166370250 25 mL Glass Bottle
AC166370050 5 mL Glass Bottle
AC166370100 10 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC166370250 Supplier Thermo Scientific Chemicals Supplier No. 166370250
Add to Cart
This item is not returnable. View return policy

CAS: 13031-32-8 | CH3NO2 | 64.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13031-32-8
Molecular Formula CH3NO2
Molecular Weight (g/mol) 64.06
MDL Number MFCD00044214
InChI Key LYGJENNIWJXYER-FIBGUPNXSA-N
Synonym nitromethane-d3, nitro 2h3 methane, nitro 2 h? methane, nitromethane-d3, 99 atom % d, methane-d3, nitro, cd3no2, trideuteronitromethane, methane-d3-, nitro, nitromethane-d3, ≥99 atom % d, contains 1 % v/v tms
PubChem CID 123293
IUPAC Name trideuterio(nitro)methane
SMILES [2H]C([2H])([2H])[N+]([O-])=O

Specifications

Physical Form Liquid
Quantity 25 mL
Color Colorless
Packaging Glass Bottle
Specific Gravity 1.18
Refractive Index 1.3795
Flash Point 35°C
Density 1.1800g/mL
Water 0.1% max. (K.F.)
Deuterium (D) 98.5% min.
Boiling Point 100.0°C
Linear Formula CD3NO2
Formula Weight 64.06
Grade NMR
Chemical Name or Material Nitromethane-d3, 99 atom % D
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Flammable liquid and vapor.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 235-892-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.