Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Nitromethanetrispropionic acid

Catalog No. 36123225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-123-225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-123-225G Supplier Sigma Aldrich Supplier No. 36123225G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Nitromethanetrispropionic acid

Specifications

CAS 59085-15-3
Melting Point 181°C to 184°C
Molecular Formula C10H15NO8
Linear Formula NO2C(CH2CH2COOH)3
Quantity 25 g
MDL Number MFCD00077684
Synonym 4-(2-Carboxyethyl)-4-nitroheptanedioic acid; 4-(2-Carboxyethyl)-4-nitropimelic acid; Tris(2-carboxyethyl)nitromethane
Molecular Weight (g/mol) 277.23
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.