Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nitromide (3,5-Dinitrobenzamide) | 121-81-3 | 98.2% | 250 MG
SDP

Catalog No. 5000406530
Encompass_Preferred

Nitromide is an anti-parasitic agent that appears as a solid with a color ranging from light yellow to yellow.

  • Anti-parasitic agent
  • Appears as a solid
  • Color: light yellow to yellow
  • Purity: 98.2%
  • Molecular weight: 211.13
  • Formula: C7H5N3O5
  • CAS number: 121-81-3

Catalog No. 50-004-06530 Supplier Medchemexpress LLC Supplier No. 250MGHYB0945
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nitromide is an anti-parasitic agent that appears as a solid with a color ranging from light yellow to yellow.

  • Anti-parasitic agent
  • Appears as a solid
  • Color: light yellow to yellow
  • Purity: 98.2%
  • Molecular weight: 211.13
  • Formula: C7H5N3O5
  • CAS number: 121-81-3

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.