Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitron, For Spectrophotometric Det. of Nitrate and Perchlorate, ≥97.0%, MilliporeSigma™ Supelco™

Catalog No. 111013572 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3572 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3572 Supplier MilliporeSigma Supelco Supplier No. 7349050G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD00005174
Synonym 1,4-Diphenyl-3-phenylamino-1,2,4-triazolium hydroxide inner salt; 3,5,6-Triphenyl-2,3,5,6-tetraazabicyclo[2.1.1 ]hex-1-ene; 4,5-Dihydro-2,4-diphenyl-5-(phenylimino)-1 H-1,2,4-triazolium hydroxide inner salt

Specifications

Melting Point 189°C to 190°C
CAS 2218-94-2
Packaging Glass Bottle
Linear Formula C20H16N4
Quantity 50 g
Formula Weight 312.37
Percent Purity ≥97.0% (NT); ≥97.0%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.