Learn More
Crescent Chemical Co Inc NITROPHENYL-B-D-GALACTOPYRANOS
SDP
Catalog No. 50247938
Encompass_Preferred
Nitrophenyl-b-D-galactopyranoside research grade Substrate for β-D-galactosidase. Size - 5G Storage Conditions - +2 °C TO +8 °C Catalog Number - 30710.02 CAS 369-07-3
Catalog No. 50-247-938 Supplier Crescent Chemical Co Inc Supplier No. SE30710.02
Nitrophenyl-b-D-galactopyranoside research grade Substrate for β-D-galactosidase. Size - 5G Storage Conditions - +2 °C TO +8 °C Catalog Number - 30710.02 CAS 369-07-3