Learn More
Crescent Chemical Co Inc NITROPHENYL PHOSPHATE-NA2-SALT
SDP
Catalog No. 50247941
Encompass_Preferred
Nitrophenyl phosphate-Na2-salt analytical grade Filled under nitrogen. High quality substrate for alkaline and acid phosphatase (1). Size - 25G Storage Conditions - -15 °C TO -25 °C Catalog Number - 30770.02 CAS 4264-83-9
Catalog No. 50-247-941 Supplier Crescent Chemical Co Inc Supplier No. SE30770.02
Nitrophenyl phosphate-Na2-salt analytical grade Filled under nitrogen. High quality substrate for alkaline and acid phosphatase (1). Size - 25G Storage Conditions - -15 °C TO -25 °C Catalog Number - 30770.02 CAS 4264-83-9