Learn More
Medchemexpress LLC Nitrosobenzene | 586-96-9 | >98.0% | 1 G
SDP
Catalog No. 5000232453
Encompass_Preferred
Nitrosobenzene (C6H5NO) is an organic nitroso compound used as a reagent and intermediate in organic synthesis and biochemical research. It functions as a radical scavenger and is used to study oxidative DNA damage and neutrophil respiratory burst reactions.
  • High purity (>98.0%) reagent grade suitable for research applications.
  • Molecular weight 107.11 g/mol and formula C6H5NO.
  • Commonly used as an intermediate for azoxybenzene and azobenzene synthesis.
  • Useful as a radical scavenger in assays of oxidative DNA damage.
  • Supplied as a 1 G solid for laboratory use; follow SDS for handling and safety.
Catalog No. 50-002-32453 Supplier Medchemexpress LLC Supplier No. HY1216411G
Nitrosobenzene (C6H5NO) is an organic nitroso compound used as a reagent and intermediate in organic synthesis and biochemical research. It functions as a radical scavenger and is used to study oxidative DNA damage and neutrophil respiratory burst reactions.
  • High purity (>98.0%) reagent grade suitable for research applications.
  • Molecular weight 107.11 g/mol and formula C6H5NO.
  • Commonly used as an intermediate for azoxybenzene and azobenzene synthesis.
  • Useful as a radical scavenger in assays of oxidative DNA damage.
  • Supplied as a 1 G solid for laboratory use; follow SDS for handling and safety.