Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nitrosobenzene | 586-96-9 | 99.0% | 500 MG
SDP

Catalog No. 5000230226
Encompass_Preferred

Nitrosobenzene is an organic nitroso compound used as a free-radical scavenger in research applications; it is commonly employed to study oxidative DNA damage and nitroso-compound-induced respiratory burst in neutrophils.

  • Used as a free-radical scavenger in oxidative stress assays.
  • Typical applications include studies of DNA oxidation and neutrophil respiratory burst.
  • CAS number 586-96-9; molecular formula C6H5NO.
  • Molecular weight 107.11 g/mol.
  • Available in small pack sizes for laboratory use (e.g., 500 MG).

Catalog No. 50-002-30226 Supplier Medchemexpress LLC Supplier No. HY121641500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nitrosobenzene is an organic nitroso compound used as a free-radical scavenger in research applications; it is commonly employed to study oxidative DNA damage and nitroso-compound-induced respiratory burst in neutrophils.

  • Used as a free-radical scavenger in oxidative stress assays.
  • Typical applications include studies of DNA oxidation and neutrophil respiratory burst.
  • CAS number 586-96-9; molecular formula C6H5NO.
  • Molecular weight 107.11 g/mol.
  • Available in small pack sizes for laboratory use (e.g., 500 MG).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.