Learn More
Electron Microscopy Sciences NitroTAG™ Cryo Barcodes 2 x 1" on 1" core, Orange
SDP
Supplier:  Electron Microscopy Sciences 7742021O
Encompass_Preferred
These cryo barcode labels for liquid nitrogen storage feature an extra-permanent adhesive for secure long-term biopreservation under cryogenic laboratory conditions, including helium, liquid, and vapor phase liquid nitrogen, freezers, and dry ice transportation. The labels are ideal for use on cryo boxes, vials, tubes, plastic bags, and plates. They are resistant to alcohol and multiple freeze-thaw cycles and no cure time allows submerging in liquid nitrogen immediately. With extra-permanent adhesives, they will stick to wet surfaces. Corners are rounded. The labels are compatible with most thermal-transfer printers.
Size: 2 x 1"
Color: Orange
Core: 1" core
Labels across: 1 rectangle
Perforated: Yes
Catalog No. 50-193-0169