Learn More
Electron Microscopy Sciences NitroTAG™ Cryo Barcodes ¼" dot on 1" core, White
SDP
Supplier:  Electron Microscopy Sciences 7742035W
Encompass_Preferred
These cryo barcode labels for liquid nitrogen storage feature an extra-permanent adhesive for secure long-term biopreservation under cryogenic laboratory conditions, including helium, liquid, and vapor phase liquid nitrogen, freezers, and dry ice transportation. The labels are ideal for use on cryo boxes, vials, tubes, plastic bags, and plates. They are resistant to alcohol and multiple freeze-thaw cycles and no cure time allows submerging in liquid nitrogen immediately. With extra-permanent adhesives, they will stick to wet surfaces. Corners are rounded. The labels are compatible with most thermal-transfer printers.
Size: ¼"
Color: White
Core: 1" core
Labels across: 2 circles
Perforated: Yes + notch
Catalog No. 50-193-0211