Learn More
Research Products International Corp NitroTag Cryogenic Barcode Labels, 1.25x0.875, 1"" Core, White, 1000 Labels
SDP
Supplier:  Research Products International Corp 247187
Encompass
NitroTag labels with superior permanent adhesive adhere to wet surfaces and can immediately be submerged into liquid nitrogen. Recommended for long-term storage and biopreservation of cryogenic vials in liquid and vapor phase nitrogen; liquid helium; -80° freezer and dry ice transportation. Labels accept all forms of thermal transfer printing and are alcohol resistant. Recommended for tubes exceeding 1.5ml in size including 2ml microcentrifuge tubes and screw-cap cryovials.
Catalog No. 50-197-8291