Learn More
AdipoGen Nitrotetrazolium blue chloride (100 mg)
SDP
Catalog No. 501490505
Encompass_Preferred
Nitrotetrazolium blue chloride (100 mg), AdipoGen Life Sciences. Chemical. CAS: 298-83-9. Formula: C40H30Cl2N10O6. MW: 817.2. Synthetic. NADPH-diaphorase substrate that competitively inhibits NOS (nitric oxide synthase). Well-known scavenger of superoxide anions. Dye that is used for detection of alkaline phosphatase in combination with 5-bromo-4-chloro-3-indoxyl phosphate (BCIP). This substrate system produces an insoluble NBT diformazan end product that is blue in color and can be observed visually. When used with BCIP, it is suitable for detection of alkaline phosphatase in western blots, for immunohistological staining procedures and for colorimetric indication of bacterial infection in blood samples. Used as a redox indicator for enzymatic reactions including dehydrogenases, threonine deaminase, glucose-6-phosphate dehydrogenase, phosphofructokinase on polyacrylamide gels, oxidases on polyacrylamide gels and pentose shunt dehydrogenses.
Catalog No. 50-149-0505 Supplier AdipoGen Supplier No. CDXN0009M100
Nitrotetrazolium blue chloride (100 mg), AdipoGen Life Sciences. Chemical. CAS: 298-83-9. Formula: C40H30Cl2N10O6. MW: 817.2. Synthetic. NADPH-diaphorase substrate that competitively inhibits NOS (nitric oxide synthase). Well-known scavenger of superoxide anions. Dye that is used for detection of alkaline phosphatase in combination with 5-bromo-4-chloro-3-indoxyl phosphate (BCIP). This substrate system produces an insoluble NBT diformazan end product that is blue in color and can be observed visually. When used with BCIP, it is suitable for detection of alkaline phosphatase in western blots, for immunohistological staining procedures and for colorimetric indication of bacterial infection in blood samples. Used as a redox indicator for enzymatic reactions including dehydrogenases, threonine deaminase, glucose-6-phosphate dehydrogenase, phosphofructokinase on polyacrylamide gels, oxidases on polyacrylamide gels and pentose shunt dehydrogenses.