Learn More
AdipoGen Nitrotetrazolium blue chloride
SDP
Catalog No. 502560443
Encompass_Preferred
Chemical. CAS 298-83-9. Formula C40H30Cl2N10O6. MW 817.2. Synthetic. NADPH-diaphorase substrate that competitively inhibits NOS nitric oxide synthase. Well-known scavenger of superoxide anions. Dye that is used for detection of alkaline phosphatase in combination with 5-bromo-4-chloro-3-indoxyl phosphate BCIP. This substrate system produces an insoluble NBT diformazan end product that is blue in color and can be observed visually. When used with BCIP, it is suitable for detection of alkaline phosphatase in western blots, for immunohistological staining procedures and for colorimetric indication of bacterial infection in blood samples. Used as a redox indicator for enzymatic reactions including dehydrogenases, threonine deaminase, glucose-6-phosphate dehydrogenase, phosphofructokinase on polyacrylamide gels, oxidases on polyacrylamide gels and pentose shunt dehydrogenses. The NBT/BCIP reaction is also used for colorimetric/spectrophotometric activity assays of oxidoreductases.
Catalog No. 50-256-0443 Supplier AdipoGen Supplier No. CDXN0009G001
Chemical. CAS 298-83-9. Formula C40H30Cl2N10O6. MW 817.2. Synthetic. NADPH-diaphorase substrate that competitively inhibits NOS nitric oxide synthase. Well-known scavenger of superoxide anions. Dye that is used for detection of alkaline phosphatase in combination with 5-bromo-4-chloro-3-indoxyl phosphate BCIP. This substrate system produces an insoluble NBT diformazan end product that is blue in color and can be observed visually. When used with BCIP, it is suitable for detection of alkaline phosphatase in western blots, for immunohistological staining procedures and for colorimetric indication of bacterial infection in blood samples. Used as a redox indicator for enzymatic reactions including dehydrogenases, threonine deaminase, glucose-6-phosphate dehydrogenase, phosphofructokinase on polyacrylamide gels, oxidases on polyacrylamide gels and pentose shunt dehydrogenses. The NBT/BCIP reaction is also used for colorimetric/spectrophotometric activity assays of oxidoreductases.