Learn More
Abcam Nitrotetrazolium blue chloride (NBT), Chromogenic substrate, 1G
Supplier:  Abcam AB1462621G
Encompass_Preferred
MW 817.64 g/mol. Chromogenic substrate for dehydrogenases and other oxidases that, like other tetrazolium compounds, can be reduced to produce a colored formazan derivative. Acts as an NADPH-diaphorase substrate that competitively inhibits nitric oxide synthase. Used for detection of alkaline phosphatase in combination with 5-bromo-4-chloro-3-indoxyl phosphate (BCIP).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4785