Learn More
Cayman Chemical NItrotyrosIn IP KIt 1ea
SDP
Catalog No. 502896382
Encompass_Preferred
For the capture and pulldown of nitrated proteins from cell and tissue lysates
Catalog No. 50-289-6382 Supplier Cayman Chemical Supplier No. 6012201 EA
For the capture and pulldown of nitrated proteins from cell and tissue lysates