Learn More
Cell Biolabs Inc OxiSelect™ Nitrotyrosine ELISA Kit, 96 assays
Supplier:  Cell Biolabs Inc STA305
Encompass_Preferred
The modification of tyrosine residues in proteins to 3-nitrotyrosine by peroxynitrite (Figure 1) or other potential nitrating agents has been detected in biological systems that are subject to oxidative stress. Detection of nitrotyrosine-containing proteins has been reported in many human and animal diseases or cellular models of disease. The OxiSelect™ Nitrotyrosine ELISA Kit is a competitive enzyme immunoassay developed for rapid detection and quantitation of 3-nitrotyrosine in protein sample. The quantity of 3-nitrotyrosine in protein sample is determined by comparing its absorbance with that of a known nitrated BSA standard curve. The kit has a nitrotyrosine detection sensitivity range of 20 nM to 8.0 μM. Each kit provides sufficient reagents to perform up to 96 assays, including standard curve and unknown protein samples.
Catalog No. NC9972742