Learn More
Enzo Life Sciences Nitrotyrosine monoclonal antibody (HM11) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLSA4680100
Encompass_Preferred
Clone: HM11. Isotype: IgG1. Immunogen: Nitrated KLH. Species reactivity: Species independent. Applications: ELISA, IHC, WB. Purity: Sterile. Formulation: Liquid. In PBS.
Catalog No. 50-201-1831