Learn More
Apexbio Technology LLC Nitroxoline,1g CAS# 4008-48-4
SDP
Supplier:  Apexbio Technology LLC B61351000
Encompass_Preferred
Nitroxoline has been clinically used since 1962 for the treatment of urinary tract infections, especially those caused by gram negative bacilli. The current renewal of nitroxoline is due to its recently found activity against fungi, U. urealyticum, Mycoplasma, and Trichomonas.In vitro: The machnistic study showed that nitroxoline, in the treatment of acute or recurrent urinary tract infections caused by Escherichia coli, could be decreased in the presence of Mg2+ and Mn2+ but not Ca2+. Moreover, with the divalent metal ions, a shift in the nitroxoline A448 indicated the formation of drug-ion complexes and a clear correlation was observed between the chelating property and antibacterial activity of nitroxolinet. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0561