Learn More
Medchemexpress LLC Nitroxoline-d4 (8-Hydroxy-5-nitroquinoline-d4) | 4008-48-4 | 1 MG
SDP
Catalog No. 5000411619
Encompass_Preferred
Nitroxoline-d4 is a deuterium-labeled form of Nitroxoline. Nitroxoline is an antibiotic effective against biofilm infections, primarily by chelating Fe2+ and Zn2+ ions from the biofilm matrix. This compound is intended for research use only and is not sold to patients. Deuterated compounds like Nitroxoline-d4 can serve as tracers for quantitation during drug development and may influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements incorporated into drug molecules for quantitation during drug development.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-11619 Supplier Medchemexpress LLC Supplier No. HYB1159S1MG
Nitroxoline-d4 is a deuterium-labeled form of Nitroxoline. Nitroxoline is an antibiotic effective against biofilm infections, primarily by chelating Fe2+ and Zn2+ ions from the biofilm matrix. This compound is intended for research use only and is not sold to patients. Deuterated compounds like Nitroxoline-d4 can serve as tracers for quantitation during drug development and may influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements incorporated into drug molecules for quantitation during drug development.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.