Learn More
Bioworld Nitsch Medium, 50 L
Catalog No. 50254753
Encompass_Preferred
Nitsch MediumContains Micronutrients and Macronutrients as described by Nitsch & Nitsch (1969) Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25 L or 50 L of medium, or as 10 units to make 1 L of mediumPlant tissue culture testedWorking Concentration: 2.10 g/LpH: 3.5−4.5Soluble in water
Catalog No. 50-254-753 Supplier Bioworld Supplier No. 306300953
Nitsch MediumContains Micronutrients and Macronutrients as described by Nitsch & Nitsch (1969) Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25 L or 50 L of medium, or as 10 units to make 1 L of mediumPlant tissue culture testedWorking Concentration: 2.10 g/LpH: 3.5−4.5Soluble in water