Learn More
Bioworld Nitsch Medium w/ Vitamins, 50 L
Catalog No. 50254755
Encompass_Preferred
Nitsch MediumContains Macronutrients, Micronutrients, and Vitaminsas described by Nitsch & Nitsch (1969)Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested Working Concentration: 2.21 g/LSoluble in water
Catalog No. 50-254-755 Supplier Bioworld Supplier No. 306300963
Nitsch MediumContains Macronutrients, Micronutrients, and Vitaminsas described by Nitsch & Nitsch (1969)Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested Working Concentration: 2.21 g/LSoluble in water