Learn More
Research Products International Corp Nitschs' Medium, Powder, Makes 5 Liters
SDP
Supplier:  Research Products International Corp N120705.0
Encompass_Preferred
Basal salt mixture containing micro and macro elements as described by Nitsch & Nitsch. (1969).
Catalog No. 50-213-534