Learn More
Research Products International Corp Nitschs' Medium with Vitamins, Powder, Makes 10 Liters
SDP
Catalog No. 50213536
Encompass_Preferred
Basal salt mixture containing micro and macro elements with vitamins as described by Nitsch & Nitsch (1969).
Catalog No. 50-213-536 Supplier Research Products International Corp Supplier No. N1209010.0
Basal salt mixture containing micro and macro elements with vitamins as described by Nitsch & Nitsch (1969).