Learn More
Research Products International Corp Nitschs Vitamin Mixture, 10.85 Grams of Powder, Makes 100 Milliliters of 1000X Solution
SDP
Catalog No. 50213563
Encompass_Preferred
Powdered vitamin mixture as described by Nitsch & Nitsch (1969). Package contains vitamins to prepare a 1000X vitamin stock solution.
Use 1ml vitamin stock solution to prepare 1 liter Nitsch medium of final vitamin concentration.
Catalog No. 50-213-563 Supplier Research Products International Corp Supplier No. N430100.1
Powdered vitamin mixture as described by Nitsch & Nitsch (1969). Package contains vitamins to prepare a 1000X vitamin stock solution.
Use 1ml vitamin stock solution to prepare 1 liter Nitsch medium of final vitamin concentration.