Learn More
Bioworld Nitsch Vitamin Mix, 100 mL
Catalog No. 50254757
Encompass_Preferred
Nitsch Vitamin MixContains Vitamins as described by Nitsch & Nitsch (1969)Package includes 10.85 g mix to make 100 mL of 1000x stock solution Use 1 ml/L of mediumContents: Biotin: 0.05 mg/LFolic acid: 0.50 mg/LGlycine: 2.00 mg/L myo Inositol: 100.0 mg/L Nicotinic acid: 5.0 mg/L Pyridoxine HCl: 0.5 mg/L Thiamine HCl: 0.5 mg/L
Catalog No. 50-254-757 Supplier Bioworld Supplier No. 306300981
Nitsch Vitamin MixContains Vitamins as described by Nitsch & Nitsch (1969)Package includes 10.85 g mix to make 100 mL of 1000x stock solution Use 1 ml/L of mediumContents: Biotin: 0.05 mg/LFolic acid: 0.50 mg/LGlycine: 2.00 mg/L myo Inositol: 100.0 mg/L Nicotinic acid: 5.0 mg/L Pyridoxine HCl: 0.5 mg/L Thiamine HCl: 0.5 mg/L