Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KINOVATE LIFE SCIENCES INC NITTOPHASE HL UNYLINKE, 50 g, 350 μmol/g

Catalog No. NC1789154
Encompass

KINOVATE LIFE SCIENCES INC NITTOPHASE HL UNYLINKE 350 50G is designed for efficient and high-quality oligonucleotide synthesis using advanced technologies. This solid support simplifies the synthesis process and ensures high yields and purity across various applications. It is compatible with standard reagents and conditions, resulting in improved product quality.

  • Product type: Solid support for oligonucleotide synthesis
  • Technology: NittoPhaseHL, UnyLinker™
  • Quantity: 50 g
  • Loading: 350 μmol/g
  • Support material: Cross-linked polystyrene bead
  • Coupling efficiency: >99%
  • End product: 3'-OH oligonucleotides
  • Compliance: ISO 9001:2015 guidelines
  • Compatible with: Standard reagents, synthesis conditions, and various chemistries
  • Applications include: Phosphodiester, phosphorothioate, 2'-O-alkyl, siRNA, LNA, and other modified oligonucleotides

Catalog No. NC1789154 Supplier KINOVATE LIFE SCIENCES INC Supplier No. 20000335050G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KINOVATE LIFE SCIENCES INC NITTOPHASE HL UNYLINKE 350 50G is designed for efficient and high-quality oligonucleotide synthesis using advanced technologies. This solid support simplifies the synthesis process and ensures high yields and purity across various applications. It is compatible with standard reagents and conditions, resulting in improved product quality.

  • Product type: Solid support for oligonucleotide synthesis
  • Technology: NittoPhaseHL, UnyLinker™
  • Quantity: 50 g
  • Loading: 350 μmol/g
  • Support material: Cross-linked polystyrene bead
  • Coupling efficiency: >99%
  • End product: 3'-OH oligonucleotides
  • Compliance: ISO 9001:2015 guidelines
  • Compatible with: Standard reagents, synthesis conditions, and various chemistries
  • Applications include: Phosphodiester, phosphorothioate, 2'-O-alkyl, siRNA, LNA, and other modified oligonucleotides

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.