Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Nitty-Gritty Soil Test Kit
SDPSCI_ED

Catalog No. S65404 Shop All LaMotte Company Products
Change view
Click to view available options
Age:
8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Age
S65404 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S65404 Supplier LaMotte™ Supplier No. 5913
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Reagents for 10 tests for each test factor, games and actvities and instruction manual

Investigate the basics of soil science and healthy plant requirements for proper soil texture, ideal levels of soil pH and nutrients.

  • Learn to properly care for plants and perform pH, nitrate, phosphate and potassium soil tests using nonhazardous TesTabs™ methods
  • Ideal demos for elementary students

Order Info

Grades 3+.

Specifications

Grade 3+
Age 8
For Use With (Application) Learn to properly care for plants and perform pH, nitrate, phosphate and potassium soil tests
Includes Reagents for ten tests for each test factor
Class Size 1 Class
Product Type Soil Macronutrients Test Kit
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.