Learn More
Medchemexpress LLC HY-P9903 5mg Medchemexpress, Nivolumab CAS:946414-94-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP99035MG
Encompass_Preferred
Medchemexpress, HY-P9903 5mg Nivolumab CAS:946414-94-4 Nivolumab is a programmed death receptor-1 ( PD-1 ) blocking antibody to treat advanced (metastatic) non-small cell lung cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4391