Learn More
TARGETMOL CHEMICALS INC NIVOLUMAB 5MG
SDP
Catalog No. 502578935
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Nivolumab anti-PD-1) is a genetically engineered fully human immunoglobulin Ig) G4 monoclonal antibody directed against the negative immunoregulatory human cell surface receptor programmed death-1PD-1PCD-1) with immune checkpoint inhibitory and antineoplastic activities. purity: 98%
Catalog No. 50-257-8935 Supplier TARGETMOL CHEMICALS INC Supplier No. T99075MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Nivolumab anti-PD-1) is a genetically engineered fully human immunoglobulin Ig) G4 monoclonal antibody directed against the negative immunoregulatory human cell surface receptor programmed death-1PD-1PCD-1) with immune checkpoint inhibitory and antineoplastic activities. purity: 98%