Learn More
Abcam Nivolumab ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB2376511X96TESTS
Encompass_Preferred
Nivolumab ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Nivolumab in Human in Plasma, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5185