Learn More
LAB BIOREAGENTS Nivolumab Elisa Kit
Supplier:  LAB BIOREAGENTS NEK011726
Encompass
Nivolumab ELISA Kit is a sandwich enzyme-linked immunoassay for the quantitative determination of Nivolumab with high sensitivity and specificity in serum and plasma.Kit PerformanceSensitivity: 5 ng/mLAssay Range: 5 – 500 ng/mLCross-reactivity: There is no cross reaction with native serum immunoglobulinsRecovery rate: 80 – 120% with normal human serum samples with known concentrations.Precision: Intra-Assay: CV < 20%; Inter-Assay: CV < 20%
Catalog No. 50-195-9287