Learn More
Medchemexpress LLC Nizatidine-d3 | 1246833-99-7 | 98.8% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0310S1MG
Encompass_Preferred
Nizatidine-d3 is the deuterium labeled Nizatidine. Nizatidine is a potent and orally active histamine H2 receptor antagonist, used for the research of stomach and intestines ulcers. It decreases the secretion of gastric acid the stomach makes and prevents ulcers from coming back after they have healed in animal models.
  • Potent and orally active histamine H2 receptor antagonist
  • Used for research of stomach and intestines ulcers
  • Decreases secretion of gastric acid
  • Prevents ulcers from returning after healing
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Targets the H2 receptor
Catalog No. 50-004-06697