Learn More
Medchemexpress LLC Nizatidine-d3 | 1246833-99-7 | 98.8% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0310S5MG
Encompass_Preferred
Nizatidine-d3 is the deuterium-labeled form of Nizatidine, a potent and orally active histamine H2 receptor antagonist. It is used in the research of stomach and intestinal ulcers by decreasing gastric acid secretion and preventing ulcer recurrence. Its deuterium labeling makes it valuable as a tracer and internal standard for quantitative analysis in drug development.
  • Deuterium labeled Nizatidine
  • Potent and orally active histamine H2 receptor antagonist
  • Used for research of stomach and intestines ulcers
  • Decreases secretion of gastric acid
  • Prevents ulcers from returning after healing in animal models
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-05467