Learn More
Medchemexpress LLC Nizatidine-d3 | 1246833-99-7 | MFCD32641299 | 98.8% | 334.48 | C12H18D3N5O2S2 | 10 MG
SDP
Catalog No. 5000296871
Encompass_Preferred
Nizatidine-d3 is the deuterium-labeled form of nizatidine supplied as an isotope-labeled reference standard for research applications, including mass spectrometry, pharmacokinetic, and metabolic studies. It is intended for use as a labeled internal standard to support quantitative analytical workflows.
  • Deuterium-labeled internal standard for mass spectrometry assays.
  • High chemical purity: 98.8%.
  • CAS number 1246833-99-7 for unambiguous identification.
  • Available in small-mass packaged quantities suitable for analytical use.
  • Useful for ADME, PK/PD, and quantitative analytical studies.
Catalog No. 50-002-96871 Supplier Medchemexpress LLC Supplier No. HYB0310S10MG
Nizatidine-d3 is the deuterium-labeled form of nizatidine supplied as an isotope-labeled reference standard for research applications, including mass spectrometry, pharmacokinetic, and metabolic studies. It is intended for use as a labeled internal standard to support quantitative analytical workflows.
  • Deuterium-labeled internal standard for mass spectrometry assays.
  • High chemical purity: 98.8%.
  • CAS number 1246833-99-7 for unambiguous identification.
  • Available in small-mass packaged quantities suitable for analytical use.
  • Useful for ADME, PK/PD, and quantitative analytical studies.