Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nizatidine 97.0+%, TCI America™
SDP

Catalog No. N09335G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09335G 5 g
N093325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09335G Supplier TCI America Supplier No. N09335G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 76963-41-2
Molecular Formula C12H21N5O2S2
Molecular Weight (g/mol) 331.453
MDL Number MFCD00865660
InChI Key SGXXNSQHWDMGGP-IZZDOVSWSA-N
PubChem CID 3033637
IUPAC Name (E)-1-N'-[2-[[2-[(dimethylamino)methyl]-1,3-thiazol-4-yl]methylsulfanyl]ethyl]-1-N-methyl-2-nitroethene-1,1-diamine
SMILES CNC(=C[N+](=O)[O-])NCCSCC1=CSC(=N1)CN(C)C

Specifications

Melting Point 132°C
Color White-Yellow
Quantity 5 g
Synonym N-[2-[[[2-[(Dimethylamino)methyl]-4-thiazolyl]methyl]thio]ethyl]-N′C-methyl-2-nitro-1,1-ethenediamine
Formula Weight 331.45
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nizatidine
RTECSNumber KM6565000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.