Learn More
Enzo Life Sciences Neurokinin-1 receptor (rat) polyclonal antibody (25µl)
SDP
Supplier:  Enzo Life Sciences BMLNA43000025
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 393-407 (K393TMTESSSFYSNMLA407) of C-terminal rat NK1r (neurokinin-1 receptor). Source: From rabbit serum. Species reactivity: Rat. Applications: IHC. Formulation: Liquid. Antiserum containing 0.1% sodium azide.
Catalog No. 50-201-1191