Learn More
ATCC NK-92; Natural Killer Cell; Human (Homo sapiens); NK-92 is a registered; trademark of Nantkwest, Inc
Catalog No. 502382372
Encompass
NK-92 cells are an interleukin-2 (IL-2) dependent natural killer cell line derived from peripheral blood mononuclear cells from a 50-year-old, White male with rapidly progressive non-Hodgkin's lymphoma. Use NK-92 cells in your cancer, immunology, and toxicology research.
Catalog No. 50-238-2372 Supplier ATCC Supplier No. CRL2407
NK-92 cells are an interleukin-2 (IL-2) dependent natural killer cell line derived from peripheral blood mononuclear cells from a 50-year-old, White male with rapidly progressive non-Hodgkin's lymphoma. Use NK-92 cells in your cancer, immunology, and toxicology research.