Learn More
ATCC NK-92MI; Natural Killer Cell; Human (Homo sapiens); NK-92 is a registered; trademark of Nantkwest, Inc
Catalog No. 502383549
Encompass
NK-92 MI are natural killer cells that are cytotoxic to a wide range of malignant cells; it kills both K562 cells and Daudi cells in chromium release assays. This cell line was derived from peripheral blood mononuclear cells from a 50-year-old, White male with rapidly progressive non-Hodgkin's lymphoma. Use this cell line in your immunology and cancer research.
Catalog No. 50-238-3549 Supplier ATCC Supplier No. CRL2408
NK-92 MI are natural killer cells that are cytotoxic to a wide range of malignant cells; it kills both K562 cells and Daudi cells in chromium release assays. This cell line was derived from peripheral blood mononuclear cells from a 50-year-old, White male with rapidly progressive non-Hodgkin's lymphoma. Use this cell line in your immunology and cancer research.