Learn More
Aviva Systems Biology Corp NKAIN4 Antibody - middle region (ARP53119_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53119_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NKAIN4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NKAIN4. Alternate names: C20orf58, FAM77A, bA261N11.2ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-60931