Learn More
Aviva Systems Biology Corp NKAPL Antibody - middle region (ARP54392_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54392_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NKAPL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NKAPL. Alternate names: C6orf194, MGC126730, MGC126734, bA424I5.1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-62969