Learn More
Aviva Systems Biology Corp NKD1 Antibody - N-terminal region (ARP41285_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41285_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NKD1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human NKD1. Alternate names: Naked1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-53897