Learn More
Aviva Systems Biology Corp NKD2 Antibody - N-terminal region (ARP41287_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41287_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NKD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NKD2. Alternate names: Naked2ApplicationWB. Species reactivity: Horse, Human, Mouse, Rat
Catalog No. 50-014-57796