Learn More
Genscript Corporation NKG2A/CD159aBiotin His Av
Catalog No. 5000057677
Encompass
NKG2 also known as NKG2A(CD159A) is a member of the killer cell lectin-like receptor family NKG2A marks a unique immune effector subset preferentially co-expressing the tissue-resident CD103 molecule but not immune checkpoint inhibitors It is expressed only in NK-cells but not in T-cells or B-cells
Catalog No. 50-000-57677 Supplier Genscript Corporation Supplier No. Z04567500
NKG2 also known as NKG2A(CD159A) is a member of the killer cell lectin-like receptor family NKG2A marks a unique immune effector subset preferentially co-expressing the tissue-resident CD103 molecule but not immune checkpoint inhibitors It is expressed only in NK-cells but not in T-cells or B-cells