Learn More
ACROBiosystems ACROBiosystems Human NKG2A&CD94 Protein, Mouse IgG2a Fc Tag
Catalog No. 501956461
Encompass_Preferred
Recombinant Protein;1MG;Human NKG2A&CD94, Mouse IgG2a Fc Tag (NC4-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 113 - Leu 233 (NKG2A) & Asp 57 - Ile 179 (CD94) (Accession # P26715-1 (NKG2A) & Q13241-1 (CD94)).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 55.5 kDa. The protein migrates as 65-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6461 Supplier ACROBiosystems Supplier No. NC4H52531MG
Recombinant Protein;1MG;Human NKG2A&CD94, Mouse IgG2a Fc Tag (NC4-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 113 - Leu 233 (NKG2A) & Asp 57 - Ile 179 (CD94) (Accession # P26715-1 (NKG2A) & Q13241-1 (CD94)).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 55.5 kDa. The protein migrates as 65-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.