Learn More
ACROBiosystems ACROBiosystems Human NKG2A / CD159a Protein, Fc Tag
Supplier:  ACROBiosystems NKAH526C100UG
Encompass_Preferred
Recombinant Protein;100UG;Human NKG2A, Fc Tag (NKA-H526c) is expressed from human 293 cells (HEK293). It contains AA Pro 94 - Leu 233 (Accession # P26715-1).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 42.5 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6655