Learn More
ACROBiosystems ACROBiosystems Human NKG2A / CD159a Protein, His Tag
Catalog No. 501956741
Encompass_Preferred
Recombinant Protein;100UG;Human NKG2A, His Tag (NKA-H5244) is expressed from human 293 cells (HEK293). It contains AA Pro 94 - Leu 233 (Accession # P26715-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 17.9 kDa. The protein migrates as 30-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6741 Supplier ACROBiosystems Supplier No. NKAH5244100UG
Recombinant Protein;100UG;Human NKG2A, His Tag (NKA-H5244) is expressed from human 293 cells (HEK293). It contains AA Pro 94 - Leu 233 (Accession # P26715-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 17.9 kDa. The protein migrates as 30-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.