Learn More
Medchemexpress LLC NKG2A Human Biotin 100ug | 100ug
SDP
Catalog No. 5000193955
Encompass_Preferred
The NKG2A protein is an important immunosuppressive receptor that forms a complex with KLRD1 on lymphocyte subsets for self-non-self discrimination It recognizes HLA-E loaded with self-peptides monitors MHC class I expression in healthy cells and promotes self-tolerance NKG2A Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived NKG2A protein expressed by HEK293 with N-His N-Avi labeled tag
Catalog No. 50-001-93955 Supplier Medchemexpress LLC Supplier No. HYP78182100UG
The NKG2A protein is an important immunosuppressive receptor that forms a complex with KLRD1 on lymphocyte subsets for self-non-self discrimination It recognizes HLA-E loaded with self-peptides monitors MHC class I expression in healthy cells and promotes self-tolerance NKG2A Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived NKG2A protein expressed by HEK293 with N-His N-Avi labeled tag