Learn More
Sino Biological Human NKG2A / CD159A / KLRC1 Protein (His Tag), Biotinylated 100ug
Supplier:  Sino Biological 13905H07H1B100
Encompass_Preferred
A DNA sequence encoding the human KLRC1 (NP_002250.1) (Pro94-Leu233) was expressed with a polyhistidine tag at the N-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6219